• USA Home
  • Search Results

  • 3methylthiophene981761644411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methylthiophene

2 Products
Image of 3-Methylthiophene
CAS Number:
Empirical Formula (Hill Notation):
 C5H6S
Molecular Weight:
 98.17
 Beilstein/REAXYS Number:
 1300
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More