• USA Home
  • Search Results

  • 3nitrobenzenesulfonylchloride2216212151711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Nitrobenzenesulfonyl chloride

1 Products
Image of 3-<WBR>Nitrobenzenesulfonyl chloride
CAS Number:
Linear Formula:
 O2NC6H4SO2Cl
Molecular Weight:
 221.62
 Beilstein/REAXYS Number:
 746542
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More