• USA Home
  • Search Results

  • 3nitrobenzylchloride1715861923811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Nitrobenzyl chloride

1 Products
Synonym:   α-Chloro-3-nitrotoluene
Image of 3-Nitrobenzyl chloride
CAS Number:
Linear Formula:
 O2NC6H4CH2Cl
Molecular Weight:
 171.58
 Beilstein/REAXYS Number:
 742794
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More