• USA Home
  • Search Results

  • 3nndimethyloctylammoniopropanesulfonateinnersalt279441517876411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-(N,N-Dimethyloctylammonio)propanesulfonate inner salt

1 Products
Synonym:   Octyl sulfobetaine, SB3-8
Image of 3-(<I>N</I>,<I>N</I>-<WBR>Dimethyloctylammonio)<WBR>propanesulfonate inner salt
CAS Number:
Linear Formula:
 CH3(CH2)7N+(CH3)2CH2CH2CH2SO3-
Molecular Weight:
 279.44
 Beilstein/REAXYS Number:
 3962489
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More