• USA Home
  • Search Results

  • 3pentyn1ol84121022910411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Pentyn-1-ol

1 Products
Image of 3-Pentyn-1-ol
CAS Number:
Linear Formula:
 CH3C≡CCH2CH2OH
Molecular Weight:
 84.12
 Beilstein/REAXYS Number:
 1735967
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
208698 3-Pentyn-1-ol, 97%,