• USA Home
  • Search Results

  • 3phenyl1propanol1361912297411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Phenyl-1-propanol

2 Products
Synonym:   3-Phenylpropyl alcohol, Hydrocinnamyl alcohol
Image of 3-Phenyl-1-propanol
CAS Number:
Linear Formula:
 C6H5(CH2)3OH
Molecular Weight:
 136.19
 Beilstein/REAXYS Number:
 1857542
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Agency/Method
Show More