• USA Home
  • Search Results

  • 3phenyl1propylamine13521203857511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Phenyl-1-propylamine

1 Products
Synonym:   γ-Aminopropylbenzene, 3-Phenylpropylamine
Image of 3-<WBR>Phenyl-<WBR>1-<WBR>propylamine
CAS Number:
Linear Formula:
 C6H5(CH2)3NH2
Molecular Weight:
 135.21
 Beilstein/REAXYS Number:
 2205526
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More