• USA Home
  • Search Results

  • 3phenyl2propyn1aminehydrochloride167643001136011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Phenyl-2-propyn-1-amine hydrochloride

1 Products
Synonym:   3-Phenylpropargylamine hydrochloride
Image of 3-<WBR>Phenyl-<WBR>2-<WBR>propyn-<WBR>1-<WBR>amine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C9H10ClN
Molecular Weight:
 167.64
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More