• USA Home
  • Search Results

  • 3phenylpyridine15520100888411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Phenylpyridine

1 Products
Synonym:   3-Phenylpyridine
Image of 3-Phenylpyridine
CAS Number:
Empirical Formula (Hill Notation):
 C11H9N
Molecular Weight:
 155.20
 EC Index Number
 213-762-6
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More