• USA Home
  • Search Results

  • 3propylidenephthalide174201736959411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Propylidenephthalide

2 Products
Synonym:   3-Propylidene-1(3H)-isobenzofuranone
Image of 3-<WBR>Propylidenephthalide
CAS Number:
Empirical Formula (Hill Notation):
 C11H10O2
Molecular Weight:
 174.20
 Council of Europe no.:
 494
 EC Number:
 FEMA Number:
 2952
 Flavis number:
 10.005
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More