• USA Home
  • Search Results

  • 3quinolinecarboxaldehyde157171366942611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Quinolinecarboxaldehyde

1 Products
Image of 3-<WBR>Quinolinecarboxaldehyde
CAS Number:
Empirical Formula (Hill Notation):
 C10H7NO
Molecular Weight:
 157.17
 Beilstein/REAXYS Number:
 113269
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More