• USA Home
  • Search Results

  • 3tertbutyl4hydroxyanisole1802412100611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-tert-Butyl-4-hydroxyanisole

3 Products
Synonym:   2-tert-Butyl-4-methoxyphenol, 3-BHA, BHA
Image of 3-<I>tert</I>-<WBR>Butyl-<WBR>4-<WBR>hydroxyanisole
CAS Number:
Empirical Formula (Hill Notation):
 C11H16O2
Molecular Weight:
 180.24
 Beilstein/REAXYS Number:
 1867499
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Special Grade
Show More