• USA Home
  • Search Results

  • 3trifluoromethylbenzaldehyde1741245489711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-(Trifluoromethyl)benzaldehyde

1 Products
Synonym:   α,α,α-Trifluoro-m-tolualdehyde
Image of 3-<WBR>(Trifluoromethyl)<WBR>benzaldehyde
CAS Number:
Linear Formula:
 CF3C6H4CHO
Molecular Weight:
 174.12
 Beilstein/REAXYS Number:
 2327537
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More