• USA Home
  • Search Results

  • 3trifluoromethylbenzoicacid1901245492211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-(Trifluoromethyl)benzoic acid

1 Products
Synonym:   α,α,α-Trifluoro-m-toluic acid, 3-Carboxybenzotrifluoride
Image of 3-<WBR>(Trifluoromethyl)<WBR>benzoic acid
CAS Number:
Linear Formula:
 CF3C6H4CO2H
Molecular Weight:
 190.12
 Beilstein/REAXYS Number:
 2049239
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More