• USA Home
  • Search Results

  • 3trifluoromethylbenzylchloride1945870529311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-(Trifluoromethyl)benzyl chloride

1 Products
Synonym:   α′-Chloro-α,α,α-trifluoro-m-xylene, 3-(Chloromethyl)benzotrifluoride
Image of 3-<WBR>(Trifluoromethyl)<WBR>benzyl chloride
CAS Number:
Linear Formula:
 CF3C6H4CH2Cl
Molecular Weight:
 194.58
 Beilstein/REAXYS Number:
 743242
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More