• USA Home
  • Search Results

  • 3trifluoromethylbenzylsulfonylchloride2586512716296311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-trifluoromethylbenzylsulfonyl chloride

1 Products
Synonym:   (3-Trifluoromethylphenyl)methanesulfonyl chloride, 3-(Trifluoromethyl)benzenemethanesulfonyl chloride
Image of 3-<WBR>trifluoromethylbenzylsulfonyl chloride
CAS Number:
Empirical Formula (Hill Notation):
 C8H6ClF3O2S
Molecular Weight:
 258.65
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More