• USA Home
  • Search Results

  • 3trimethoxysilylpropylmethacrylate24835253085011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-(Trimethoxysilyl)propyl methacrylate

2 Products
Synonym:   Silane A174, [3-(Methacryloyloxy)propyl]trimethoxysilane
Image of 3-<WBR>(Trimethoxysilyl)<WBR>propyl methacrylate
CAS Number:
Linear Formula:
 H2C=C(CH3)CO2(CH2)3Si(OCH3)3
Molecular Weight:
 248.35
 Beilstein/REAXYS Number:
 1952435
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More