• USA Home
  • Search Results

  • 3trimethylsilylpropargylalcohol12824527236611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-(Trimethylsilyl)propargyl alcohol

1 Products
Synonym:   3-(Trimethylsilyl)-2-propyn-1-ol
Image of 3-<WBR>(Trimethylsilyl)<WBR>propargyl alcohol
CAS Number:
Linear Formula:
 (CH3)3SiC≡CCH2OH
Molecular Weight:
 128.24
 Beilstein/REAXYS Number:
 1902505
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More