• USA Home
  • Search Results

  • 3ureidopropionicacid1321246288411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Ureidopropionic acid

1 Products
Synonym:   N-Carbamoyl-β-alanine
Image of 3-Ureidopropionic acid
CAS Number:
Linear Formula:
 NH2CONHCH2CH2COOH
Molecular Weight:
 132.12
 Beilstein/REAXYS Number:
 1705263
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More