• USA Home
  • Search Results

  • 42aminoethylmorpholine13019203803111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(2-Aminoethyl)morpholine

1 Products
Synonym:   2-Morpholinoethylamine
Image of 4-<WBR>(2-<WBR>Aminoethyl)<WBR>morpholine
CAS Number:
Empirical Formula (Hill Notation):
 C6H14N2O
Molecular Weight:
 130.19
 Beilstein/REAXYS Number:
 104378
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More