• USA Home
  • Search Results

  • 42aminoethylpyridine122171325863411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(2-Aminoethyl)pyridine

1 Products
Image of 4-<WBR>(2-<WBR>Aminoethyl)<WBR>pyridine
CAS Number:
Empirical Formula (Hill Notation):
 C7H10N2
Molecular Weight:
 122.17
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More