• USA Home
  • Search Results

  • 43oxobutylphenylacetate20624357206311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(3-Oxobutyl)phenyl acetate

1 Products
Synonym:   4-(4-Acetoxyphenyl)-2-butanone
Image of 4-(3-Oxobutyl)phenyl acetate
CAS Number:
Linear Formula:
 CH3CO2C6H4CH2CH2COCH3
Molecular Weight:
 206.24
 Beilstein/REAXYS Number:
 1961620
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More