• USA Home
  • Search Results

  • 4413phenylenediisopropylidenebisphenol346461359525011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-(1,3-Phenylenediisopropylidene)bisphenol

1 Products
Synonym:   Bisphenol M
Image of 4,4′-(1,3-Phenylenediisopropylidene)bisphenol
CAS Number:
Linear Formula:
 C6H4[C(CH3)2C6H4OH]2
Molecular Weight:
 346.46
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More