• USA Home
  • Search Results

  • 4444porphine5101520tetrayltetrakisbenzoicacid790771460954211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid)

1 Products
Synonym:   meso-Tetraphenylporphine-4,4′,4″,4″′-tetracarboxylic acid
Image of 4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid)
CAS Number:
Empirical Formula (Hill Notation):
 C48H30N4O8
Molecular Weight:
 790.77
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More