• USA Home
  • Search Results

  • 44bipiperidyldihydrochloride241207861984811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-Bipiperidyl dihydrochloride

1 Products
Synonym:   4,4′-Bipiperidine dihydrochloride
Image of 4,4′-Bipiperidyl dihydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C10H20N2 · 2HCl
Molecular Weight:
 241.20
 Beilstein/REAXYS Number:
 3687878
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More