• USA Home
  • Search Results

  • 44cyclohexylidenebisphenol2683584355011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-Cyclohexylidenebisphenol

1 Products
Synonym:   Bisphenol Z
Image of 4,4′-Cyclohexylidenebisphenol
CAS Number:
Linear Formula:
 C6H10(C6H4OH)2
Molecular Weight:
 268.35
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More