• USA Home
  • Search Results

  • 44diaminodiphenylmethane1982610177911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-Diaminodiphenylmethane

3 Products
Synonym:   4,4′-Methylenedianiline, MDA
Image of 4,4′-Diaminodiphenylmethane
CAS Number:
Empirical Formula (Hill Notation):
 C13H14N2
Molecular Weight:
 198.26
 Beilstein/REAXYS Number:
 474706
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More