• USA Home
  • Search Results

  • 44dihydroxybiphenyl186219288611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-Dihydroxybiphenyl

1 Products
Synonym:   4,4′-Biphenol, 4,4′-Biphenyldiol, 4,4′-Diphenol
Image of 4,4′-Dihydroxybiphenyl
CAS Number:
Linear Formula:
 HOC6H4C6H4OH
Molecular Weight:
 186.21
 Beilstein/REAXYS Number:
 1908886
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More