• USA Home
  • Search Results

  • 44dimethylbenzophenone2102761197211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-Dimethylbenzophenone

1 Products
Image of 4,4′-Dimethylbenzophenone
CAS Number:
Linear Formula:
 (CH3C6H4)2CO
Molecular Weight:
 210.27
 Beilstein/REAXYS Number:
 1240527
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More