• USA Home
  • Search Results

  • 44dipyridyl1561855326411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′−Dipyridyl

1 Products
Synonym:   4,4′-Bipyridine, 4,4′-Bipyridyl
Image of 4,4′−Dipyridyl
CAS Number:
Empirical Formula (Hill Notation):
 C10H8N2
Molecular Weight:
 156.18
 Beilstein/REAXYS Number:
 113176
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
289426 4,4′−Dipyridyl, 98%,