• USA Home
  • Search Results

  • 44hydroxyphenyl2butanone16420547151211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(4-Hydroxyphenyl)-2-butanone

5 Products
Synonym:   Frambione, NSC 26515, Raspberry ketone
Image of 4-<WBR>(4-<WBR>Hydroxyphenyl)<WBR>-<WBR>2-<WBR>butanone
CAS Number:
Linear Formula:
 HOC6H4CH2CH2COCH3
Molecular Weight:
 164.20
 Beilstein/REAXYS Number:
 776080
 Council of Europe no.:
 755
 EC Number:
 FEMA Number:
 2588
 Flavis number:
 7.055
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Shelf Life
  Allergen
  Organoleptic property
  Application(s)
  Assay
  Agency/Method
Show More