• USA Home
  • Search Results

  • 44mdi2502510168811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-MDI

1 Products
Synonym:   4,4′-MDI, 4,4′-Methylenebis(phenyl isocyanate), Bis(4-isocyanatophenyl)methane
Image of 4,4′-MDI
CAS Number:
Linear Formula:
 CH2(C6H4NCO)2
Molecular Weight:
 250.25
 Beilstein/REAXYS Number:
 797662
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More