• USA Home
  • Search Results

  • 44methylenebis2methylaniline2263283888011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-Methylene-bis(2-methylaniline)

1 Products
Synonym:   4,4′-Diamino-3,3′-dimethyldiphenylmethane, 4,4′-Methylene-di(o-toluidine)
Image of 4,4′-Methylene-bis(2-methylaniline)
CAS Number:
Empirical Formula (Hill Notation):
 C15H18N2
Molecular Weight:
 226.32
 Beilstein/REAXYS Number:
 398069
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More