• USA Home
  • Search Results

  • 44nitrobenzylpyridine21422108348311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(4-Nitrobenzyl)pyridine

3 Products
Image of 4-<WBR>(4-<WBR>Nitrobenzyl)<WBR>pyridine
CAS Number:
Empirical Formula (Hill Notation):
 C12H10N2O2
Molecular Weight:
 214.22
 Beilstein/REAXYS Number:
 170877
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More