• USA Home
  • Search Results

  • 44oxydianiline2002410180411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-Oxydianiline

3 Products
Synonym:   4,4′-Diaminodiphenyl ether, 4-Aminophenyl ether
Image of 4,4′-Oxydianiline
CAS Number:
Linear Formula:
 O(C6H4NH2)2
Molecular Weight:
 200.24
 Beilstein/REAXYS Number:
 475735
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More