• USA Home
  • Search Results

  • 44thiobis2tertbutyl5methylphenol358549669511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-Thiobis(2-tert-butyl-5-methylphenol)

2 Products
Synonym:   4,4′-Thiobis(2-tert-butyl-5-methylphenol), Bis(3-tert-butyl-4-hydroxy-6-methylphenyl)sulfide
Image of 4,4′-Thiobis(2-tert-butyl-5-methylphenol)
CAS Number:
Linear Formula:
 (2-(CH3)-4-(OH)-5-((CH3)3C)C6H2)2S
Molecular Weight:
 358.54
 EC Index Number
 202-525-2
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More