• USA Home
  • Search Results

  • 4567tetrahydroindole121181361891211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,5,6,7-Tetrahydroindole

1 Products
Synonym:   2,3-Tetramethylenepyrrole, Cyclohex[b]pyrrole, NSC 122455
Image of 4,5,6,7-<WBR>Tetrahydroindole
CAS Number:
Empirical Formula (Hill Notation):
 C8H11N
Molecular Weight:
 121.18
 Beilstein/REAXYS Number:
 108853
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More