• USA Home
  • Search Results

  • 45dimethyl3hydroxy25dihydrofuran2one128132866435911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one

2 Products
Synonym:   Caramel furanone, Sotolon, Sugar lactone
Image of 4,5-<WBR>Dimethyl-<WBR>3-<WBR>hydroxy-<WBR>2,5-<WBR>dihydrofuran-<WBR>2-<WBR>one
CAS Number:
Empirical Formula (Hill Notation):
 C6H8O3
Molecular Weight:
 128.13
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Assay
  Special Grade
Show More