• USA Home
  • Search Results

  • 46diamidino2phenylindoledihydrochloride350252871890311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4′,6-Diamidino-2-phenylindole dihydrochloride

2 Products
Synonym:   2-(4-Amidinophenyl)-6-indolecarbamidine dihydrochloride, DAPI dihydrochloride
Image of 4′,6-Diamidino-2-phenylindole dihydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C16H15N5 · 2HCl
Molecular Weight:
 350.25
 Beilstein/REAXYS Number:
 4894417
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More