• USA Home
  • Search Results

  • 4713162124hexaoxa110diazabicyclo888hexacosane376492397809811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane

1 Products
Synonym:   Cryptand 222, Kryptofix® 222
Image of 4,7,13,16,21,24-<WBR>Hexaoxa-<WBR>1,10-<WBR>diazabicyclo[8.8.8]<WBR>hexacosane
CAS Number:
Empirical Formula (Hill Notation):
 C18H36N2O6
Molecular Weight:
 376.49
 Beilstein/REAXYS Number:
 620282
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More