• USA Home
  • Search Results

  • 4acetamidobenzaldehyde1631712285011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Acetamidobenzaldehyde

1 Products
Synonym:   4′-Formylacetanilide
Image of 4-<WBR>Acetamidobenzaldehyde
CAS Number:
Linear Formula:
 CH3CONHC6H4CHO
Molecular Weight:
 163.17
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More