• USA Home
  • Search Results

  • 4acetamidobenzenesulfonylazide24024215814711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Acetamidobenzenesulfonyl azide

1 Products
Synonym:   p-ABSA
Image of 4-<WBR>Acetamidobenzenesulfonyl azide
CAS Number:
Linear Formula:
 CH3CONHC6H4SO2N3
Molecular Weight:
 240.24
 Beilstein/REAXYS Number:
 2219568
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More