• USA Home
  • Search Results

  • 4acetamidotempofreeradical213301469189511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Acetamido-TEMPO, free radical

1 Products
Synonym:   4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl
Image of 4-Acetamido-TEMPO, free radical
CAS Number:
Empirical Formula (Hill Notation):
 C11H21N2O2
Molecular Weight:
 213.30
 Beilstein/REAXYS Number:
 3546225
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More