• USA Home
  • Search Results

  • 4allyl12dimethoxybenzene178239315211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Allyl-1,2-dimethoxybenzene

1 Products
Synonym:   4-Allyl-1,2-dimethoxybenzene, Eugenol methyl ether, Eugenyl methyl ether
Image of 4-<WBR>Allyl-<WBR>1,2-<WBR>dimethoxybenzene
CAS Number:
Linear Formula:
 H2C=CHCH2C6H3(OCH3)2
Molecular Weight:
 178.23
 Beilstein/REAXYS Number:
 1910871
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More