• USA Home
  • Search Results

  • 4amino2266tetramethylpiperidine156273676862411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Amino-2,2,6,6-tetramethylpiperidine

2 Products
Synonym:   Triacetonediamine, TAD
Image of 4-<WBR>Amino-<WBR>2,2,6,6-<WBR>tetramethylpiperidine
CAS Number:
Empirical Formula (Hill Notation):
 C9H20N2
Molecular Weight:
 156.27
 EC Index Number
 253-197-2
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More