• USA Home
  • Search Results

  • 4aminoantipyrine203248307811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Aminoantipyrine

4 Products
Synonym:   4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone
Image of 4-Aminoantipyrine
CAS Number:
Empirical Formula (Hill Notation):
 C11H13N3O
Molecular Weight:
 203.24
 Beilstein/REAXYS Number:
 181635
 EC Number:
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Loss
  Solubility
  Melting Point
  Special Grade
  Application(s)
Show More