• USA Home
  • Search Results

  • 4aminobenzoicacid1371415013011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Aminobenzoic acid

6 Products
Synonym:   PABA
Image of 4-Aminobenzoic acid
CAS Number:
Empirical Formula (Hill Notation):
 C7H7NO2
Molecular Weight:
 137.14
 EC Index Number
 205-753-0
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Melting Point
  Special Grade
Show More