• USA Home
  • Search Results

  • 4aminosalicylicacid153146549611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Aminosalicylic acid

2 Products
Synonym:   4-Amino-2-hydroxybenzoic acid, PAS
Image of 4-Aminosalicylic acid
CAS Number:
Linear Formula:
 H2NC6H3-2-(OH)CO2H
Molecular Weight:
 153.14
 Beilstein Registry Number:
 473071
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More