• USA Home
  • Search Results

  • 4aminotempofreeradical171261469188411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Amino-TEMPO, free radical

1 Products
Synonym:   4-Amino-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-Amino-2,2,6,6-tetramethylpiperidinyloxy, free radical, 4-Amino-TEMPO
Image of 4-Amino-TEMPO, free radical
CAS Number:
Empirical Formula (Hill Notation):
 C9H19N2O
Molecular Weight:
 171.26
 Beilstein/REAXYS Number:
 3933966
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More