• USA Home
  • Search Results

  • 4aminothiophenol12519119302811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Aminothiophenol

1 Products
Synonym:   4-Aminobenzenethiol, 4-Aminophenyl mercaptan, 4-Mercaptoaniline
Image of 4-Aminothiophenol
CAS Number:
Linear Formula:
 H2NC6H4SH
Molecular Weight:
 125.19
 Beilstein/REAXYS Number:
 906904
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More